Få mere at vide
1,2,3,4-Tetra-O-acetyl-Alpha-L-fucopyranose, TRC
Artikelnummer. 30427320
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427320 10 g
30412629 1 g
30426382 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427320 Leverandør Toronto Research Chemicals Leverandørnr. TRCT27925010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64913-16-2
Molekylær formel C14 H20 O9
Molekylvægt (g/mol) 332.3
Synonym 1,2,3,4-Tetra-O-Acetyl-α-L-Fucopyranose
IUPAC navn [(2S,3R,4R,5S,6S)-4,5,6-triacetyloxy-2-methyloxan-3-yl] acetate
SMIL C[C@@H]1O[C@@H](OC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@@H]1OC(=O)C
Tekniske data
Kemisk navn eller materiale 1,2,3,4-Tetra-O-acetyl-a-L-fucopyranose
Mængde 10 g
InChI formel InChI=1S/C14H20O9/c1-6-11(20-7(2)15)12(21-8(3)16)13(22-9(4)17)14(19-6)23-10(5)18/h6,11-14H,1-5H3/t6-,11+,12+,13-,14-/m0/s1
Formel vægt 332.1107