Få mere at vide
1,2,3,4,7,8,9-Heptachlorodibenzofuran, TRC
Artikelnummer. 30405110
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405110 1 mg
30403949 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30405110 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2650851MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55673-89-7
Molekylær formel C12 H Cl7 O
Molekylvægt (g/mol) 409.31
Synonym Dibenzofuran, 1,2,3,4,7,8,9-heptachloro-, 1,2,3,4,7,8,9-Heptachlorodibenzofuran, 1,2,3,4,7,8,9-HpCDF, 1234789-HpCDF, F 134, PCDF 134, 1,2,3,4,7,8,9-HeptaCDF, 1234789-HeptaCDF, 1234789-Heptachlorodibenzofuran
IUPAC navn 1,2,3,4,7,8,9-heptachlorodibenzofuran
SMIL Clc1cc2oc3c(Cl)c(Cl)c(Cl)c(Cl)c3c2c(Cl)c1Cl
Tekniske data
Kemisk navn eller materiale 1,2,3,4,7,8,9-Heptachlorodibenzofuran
Mængde 1 mg
InChI formel InChI=1S/C12HCl7O/c13-2-1-3-4(7(15)6(2)14)5-8(16)9(17)10(18)11(19)12(5)20-3/h1H
Formel vægt 405.7847