Få mere at vide
1,2,3,4,6-Pentachloronaphthalene, TRC
Artikelnummer. 30423566
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30423566 25 mg -
30428674 2.5 mg 2.5mg
30426017 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423566 Leverandør Toronto Research Chemicals Leverandørnr. TRCP23753025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67922-26-3
Molekylær formel C10 H3 Cl5
Molekylvægt (g/mol) 300.4
Synonym Naphthalene, 1,2,3,4,6-pentachloro-, 1,2,3,4,6-Pentachloronaphthalene, PCN 50, 1,2,3,4,6-PentaCN, 1,2,3,4,6-PCN, 12346-Pentachloronaphthalene, 12346-PentaCN, 12346-PCN, PCN-50, PCN No. 50
IUPAC navn 1,2,3,4,6-pentachloronaphthalene
SMIL Clc1ccc2c(Cl)c(Cl)c(Cl)c(Cl)c2c1
Tekniske data
Kemisk navn eller materiale 1,2,3,4,6-Pentachloronaphthalene
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C10H3Cl5/c11-4-1-2-5-6(3-4)8(13)10(15)9(14)7(5)12/h1-3H
Formel vægt 297.8677
Procent renhed >95%