Få mere at vide
1-(2,2-Difluoro-1,3-benzodioxol-5-yl)cyclopropane-2,2,3,3-d4-carboxylic Acid, TRC
Artikelnummer. 30374829
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30374829 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374829 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4520962.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(2,2-Difluoro-1,3-benzodioxol-5-yl)cyclopropane-2,2,3,3-d4-carboxylic Acid
CAS 1961281-89-9
Molekylær formel C11H4D4F2O4
Mængde 2.5 mg
InChI formel InChI=1S/C11H8F2O4/c12-11(13)16-7-2-1-6(5-8(7)17-11)10(3-4-10)9(14)15/h1-2,5H,3-4H2,(H,14,15)/i3D2,4D2
SMIL [2H]C1([2H])C([2H])([2H])C1(C(=O)O)c2ccc3OC(F)(F)Oc3c2
IUPAC navn 2,2,3,3-tetradeuterio-1-(2,2-difluoro-1,3-benzodioxol-5-yl)cyclopropane-1-carboxylic acid
Molekylvægt (g/mol) 246.2
Formel vægt 246.0642