Få mere at vide
1,12-Dodecanedioic-d20 Acid, CDN
Artikelnummer. 30311506
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311506 0.25 g -
30325453 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311506 Leverandør CDN Leverandørnr. CDND25370.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dodecanedioic-d20 Acid
CAS 89613-32-1
Molekylær formel C12 D20 H2 O4
Mængde 0.25 g
Synonym 1,10-Decanedicarboxylic Acid-d20, 1,10-Dicarboxydecane-d20, 1,12-Dodecanedioic Acid-d20, Corfree M 2-d20, Decamethylenedicarboxylic Acid-d20, NSC 400242-d20, SL-AH-d20, n-Dodecane-α,ω-dioic Acid-d20, n-Dodecanedioic Acid-d20
InChI formel InChI=1 S/C12H22O4/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16/h1-10H2,(H,13,14)(H,15,16)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2
SMIL [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosadeuteriododecanedioic acid
Molekylvægt (g/mol) 250.42
Formel vægt 250.2773 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity