missing translation for 'onlineSavingsMsg'
Få mere at vide

1,10-Decanedioic-d16 Acid, TRC

Artikelnummer. 30425812
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425812 10 mg
30425811 50 mg
30424278 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425812 Leverandør Toronto Research Chemicals Leverandørnr. TRCS21141210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 73351-71-0
Molekylær formel C10 D16 H2 O4
Molekylvægt (g/mol) 218.35
Synonym Decanedioic-d16 acid (9CI)
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecadeuteriodecanedioic acid
SMIL [2H]C([2H])(C(=O)O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O

Tekniske data

Kemisk navn eller materiale 1,10-Decanedioic-d16 Acid
Mængde 10 mg
InChI formel InChI=1S/C10H18O4/c11-9(12)7-5-3-1-2-4-6-8-10(13)14/h1-8H2,(H,11,12)(H,13,14)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2
Formel vægt 218.2209
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.