Få mere at vide
1,10-Decanedioic-d16 Acid, CDN
Artikelnummer. 30318908
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318908 0.5 g 0.5g
30310091 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318908 Leverandør CDN Leverandørnr. CDND34490.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,10-Decanedioic-d16 Acid
CAS 73351-71-0
Molekylær formel C10 D16 H2 O4
Mængde 0.5 g
Synonym Decanedioic-d16 acid (9 CI)
InChI formel InChI=1 S/C10H18O4/c11-9(12)7-5-3-1-2-4-6-8-10(13)14/h1-8H2,(H,11,12)(H,13,14)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecadeuteriodecanedioic acid
Molekylvægt (g/mol) 218.35
Formel vægt 218.2209 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity