Få mere at vide
1,1'-Sulfonylbis[2-(methylthio)ethane], TRC
Artikelnummer. 30138759
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138759 1 g
30135945 250 mg
30003286 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138759 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6994751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 137371-98-3
Molekylær formel C6H14O2S3
Molekylvægt (g/mol) 214.37
Synonym (Sulfonylbis(ethane-2,1-diyl))bis(methylsulfane)
IUPAC navn 1-methylsulfanyl-2-(2-methylsulfanylethylsulfonyl)ethane
SMIL O=S(CCSC)(CCSC)=O
Tekniske data
Kemisk navn eller materiale 1,1'-Sulfonylbis[2-(methylthio)ethane]
Mængde 1 g
InChI formel InChI=1S/C6H14O2S3/c1-9-3-5-11(7,8)6-4-10-2/h3-6H2,1-2H3
Formel vægt 214.0156