Få mere at vide
1,1'-[Propyliminobis[(2-hydroxypropane-3,1-diyl)oxy-2,1-phenylene]]bis(3-phenylpropan-1-one), Mikromol™
Artikelnummer. 17900526
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17900526 25 mg
1 options
Artikelnummer. 17900526 Leverandør Mikromol Leverandørnr. MM0405.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1'-[Propyliminobis[(2-hydroxypropane-3,1-diyl)oxy-2,1-phenylene]]bis(3-phenylpropan-1-one)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propafenone Dimer Impurity (Mixture of diastereomers)
CAS 1346603-80-2
Analyt- eller komponentnavne Propafenone Dimer Impurity (Mixture of diastereomers)
Forsendelsestilstand Room Temperature
Molekylær formel C39H45NO6
Certificeringer/compliance ISO 17025
Synonym 1,1’-[Propyliminobis[(2-hydroxypropane-1,3-diyl)oxy-2,1-phenylene]]bis(3-phenylpropan-1-one), Propafenone EP Impurity G, Propafenone BP Related Compound G
SMIL O=C(CCC1=CC=CC=C1)C2=CC=CC=C2OCC(O)CN(CCC)CC(O)COC3=C(C(CCC4=CC=CC=C4)=O)C=CC=C3
Molekylvægt (g/mol) 623.78