Få mere at vide
1,1’-Diphenyl-D10, TRC
Artikelnummer. 30369906
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30369906 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369906 Leverandør Toronto Research Chemicals Leverandørnr. TRCD486982500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Biphenyl D10
CAS 1486-01-7
Molekylær formel C12 2H10
Mængde 500 mg
Synonym Biphenyl-d10 (6CI,7CI,8CI), 1,1'-Biphenyl-d10, Perdeuteriobiphenyl, Perdeuterobiphenyl
InChI formel InChI=1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c([2H])c([2H])c2[2H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(2,3,4,5,6-pentadeuteriophenyl)benzene
Molekylvægt (g/mol) 164.27
Formel vægt 164.141