Få mere at vide
1,1-Dimethylhydrazine-d8 DCl, CDN
Artikelnummer. 30324792
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324792 0.05 g
30320023 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324792 Leverandør CDN Leverandørnr. CDND66000.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylhydrazine-D8 Monodeuterochloride
CAS 1219802-85-3
Molekylær formel C2 D8 N2 . Cl D
Mængde 0.05 g
Synonym 1,1-Dimethylhydrazine-d8 DCl, 1,1-Di(methyl-d3)hydrazine-d2 deuterochloride, 1,1-Dimethylhydrazine-d8 monodeuterochloride, N,N-Di(methyl-d3)hydrazine-d2 deuterochloride, N,N-Dimethylhydrazine-D8 monodeuterochloride
InChI formel InChI=1 S/C2H8N2.ClH/c1-4(2)3;/h3H2,1-2H3;1 H/i1D3,2D3;/hD3
SMIL [2 H]Cl.[2 H]N([2 H])N(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn (2 H)chlorane;1,1-dideuterio-2,2-bis(trideuteriomethyl)hydrazine
Molekylvægt (g/mol) 105.6147
Formel vægt 105.1019 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity