Få mere at vide
1,1-Dimethyl-3-cyanoguanidine, Mikromol™
Artikelnummer. 30458741
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458741 4 x 25 mg
30052251 100 mg
30049273 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458741 Leverandør Mikromol Leverandørnr. MM0020.1000254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1-Dimethyl-3-cyanoguanidine, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1-Dimethyl-3-cyanoguanidine
CAS 1609-06-9
Analyt- eller komponentnavne 1,1-Dimethyl-3-cyanoguanidine
Forsendelsestilstand Room Temperature
Molekylær formel C4 H8 N4
Certificeringer/compliance ISO 17025
Synonym 1,1-Dimethyl-3-cyanoguanidine
InChI formel InChI=1S/C4H8N4/c1-8(2)4(6)7-3-5/h1-2H3,(H2,6,7)
Løsningstype Neat
SMIL CN(C)C(=N)NC#N
IUPAC navn 3-cyano-1,1-dimethylguanidine
Molekylvægt (g/mol) 112.13
Mængde 4 x 25 mg
Formel vægt 112.0749 g/mol