Få mere at vide
1,1-Dimethyl-2-(methylamino)ethyl Methyl (4RS)-2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™
Artikelnummer. 17960636
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17960636 25 mg
17941336 100 mg
30459344 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17960636 Leverandør Mikromol Leverandørnr. MM3441.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1-Dimethyl-2-(methylamino)ethyl Methyl (4RS)-2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1-Dimethyl-2-(methylamino)ethyl Methyl (4RS)-2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
CAS 887769-40-6
Analyt- eller komponentnavne 1,1-Dimethyl-2-(methylamino)ethyl Methyl (4RS)-2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C21 H27 N3 O6
Certificeringer/compliance ISO 17025
Synonym 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-Pyridinedicarboxylic acid 3-[1,1-dimethyl-2-(methylamino)ethyl] 5-methyl ester, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 1,1-dimethyl-2-(methylamino)ethyl methyl ester (9CI)
InChI formel InChI=1S/C21H27N3O6/c1-12-16(19(25)29-6)18(14-8-7-9-15(10-14)24(27)28)17(13(2)23-12)20(26)30-21(3,4)11-22-5/h7-10,18,22-23H,11H2,1-6H3
SMIL CNCC(C)(C)OC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC)C
IUPAC navn 3-O-methyl 5-O-[2-methyl-1-(methylamino)propan-2-yl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 417.46
Mængde 25 mg
Formel vægt 417.19 g/mol