missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diethyl-2,2'-cyanine iodide, 97%, Thermo Scientific Chemicals

Catalog Number p-7036689
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
977-96-8
C23H23IN2
454.355
MFCD00011971
GMYRVMSXMHEDTL-UHFFFAOYSA-M
decynium 22
71299759
CHEBI:37993
(2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]
This item is not returnable. View return policy
977-96-8
C23H23IN2
454.355
MFCD00011971
GMYRVMSXMHEDTL-UHFFFAOYSA-M
decynium 22
71299759
CHEBI:37993
(2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]

1,1'-Diethyl-2,2'-cyanine iodide is used to inhibit 1-methyl-4-phenylpyridinium (MPP+), toxic metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine that induces apoptosis in cerebellar granule neurons (CGNs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Diethyl-2,2′-cyanine iodide is used to inhibit 1-methyl-4-phenylpyridinium (MPP+), toxic metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine that induces apoptosis in cerebellar granule neurons (CGNs).

Solubility
Soluble to 10 mM in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY
Melting Point 273°C (decomposition)
Quantity 250 mg
UN Number UN2811
Sensitivity Light sensitive
Solubility Information Soluble to 10 mM in dimethyl sulfoxide.
Formula Weight 454.35
Percent Purity 97%
Chemical Name or Material 1,1'-Diethyl-2,2'-cyanine iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'