Få mere at vide
1,1-Dichloroethane (Stabilized with Nitromethane, ~0.5%) (>90%), TRC
Artikelnummer. 30133420
Skift visning
Mængde:
1 g
2.5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133420 10 g
30134815 2.5 g
30010850 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133420 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43424510G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75-34-3
Molekylær formel C2 H4 Cl2
Molekylvægt (g/mol) 98.96
Synonym 1,1-Dichloroethane, 1,1-Ethylene dichloride, Dichloromethylmethane, Ethylidene chloride, Ethylidene dichloride, F 150a, HCC 150a
IUPAC navn 1,1-dichloroethane
SMIL CC(Cl)Cl
Tekniske data
Kemisk navn eller materiale 1,1-Dichloroethane
Mængde 10 g
InChI formel InChI=1S/C2H4Cl2/c1-2(3)4/h2H,1H3
Formel vægt 97.969