Learn More
1,1-Cyclopropanedicarboxylic acid dimethyl ester, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 6914-71-2 |
| Molecular Formula | C7H10O4 |
| MDL Number | MFCD00192079 |
| Quantity | 500 g |
| SMILES | COC(=O)C1(CC1)C(=O)OC |
| Molecular Weight (g/mol) | 158.15 |
| Percent Purity | 95% |
| Chemical Name or Material | 1,1-Cyclopropanedicarboxylic acid dimethyl ester |