Få mere at vide
1,1-Cyclohexanediacetic Acid Monoamide, TRC
Artikelnummer. 30137509
Skift visning
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137509 10 g
30133380 25 g
30005600 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137509 Leverandør Toronto Research Chemicals Leverandørnr. TRCC99151010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 99189-60-3
Molekylær formel C10 H17 N O3
Molekylvægt (g/mol) 199.247
Synonym Cyclohexaneacetic acid, 1-(2-amino-2-oxoethyl)-, Cyclohexaneacetic acid, 1-(carbamoylmethyl)- (6CI), 1-(2-Amino-2-oxoethyl)cyclohexaneacetic acid, 1,1-Cyclohexanediacetic acid monoamide, 1-(Carboxymethyl)cyclohexane-1-acetamide, 1-[(Aminocarbonyl)methyl]cyclohexaneacetic acid
IUPAC navn 2-[1-(2-amino-2-oxoethyl)cyclohexyl]acetic acid
SMIL NC(=O)CC1(CC(=O)O)CCCCC1
Tekniske data
Kemisk navn eller materiale 1,1-Cyclohexanediacetic Acid Monoamide
Mængde 10 g
InChI formel InChI=1S/C10H17NO3/c11-8(12)6-10(7-9(13)14)4-2-1-3-5-10/h1-7H2,(H2,11,12)(H,13,14)
Formel vægt 199.1208