Få mere at vide
1-(1-Chloromethyl-ethenyl)-2,2,4,4-tetramethylcyclohexane, TRC
Artikelnummer. 30133876
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133876 2.5 mg 2.5mg
30003780 5 mg -
30011487 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133876 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4277462.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63216-72-8
Molekylær formel C13 H23 Cl
Molekylvægt (g/mol) 214.77
Synonym 2-(3-Chloroprop-1-en-2-yl)-1,1,5,5-tetramethylcyclohexane, Rubber Oligomer
IUPAC navn 2-[1-(chloromethyl)vinyl]-1,1,5,5-tetramethyl-cyclohexane
SMIL CC1(C)CCC(C(=C)CCl)C(C)(C)C1
Tekniske data
Kemisk navn eller materiale 1-(1-Chloromethyl-ethenyl)-2,2,4,4-tetramethylcyclohexane
Mængde 2.5 mg
InChI formel InChI=1S/C13H23Cl/c1-10(8-14)11-6-7-12(2,3)9-13(11,4)5/h11H,1,6-9H2,2-5H3
Formel vægt 214.1488