Få mere at vide
1,1'-Carbonyldiimidazole-d6, CDN
Artikelnummer. 30321730
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321730 0.05 g -
30321416 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321730 Leverandør CDN Leverandørnr. CDND81870.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1'-Carbonyldiimidazole-D6
CAS 2363787-46-4
Molekylær formel C7 D6 N4 O
Mængde 0.05 g
Synonym Methanone, di(1 H-imidazol-1-yl-2,4,5-d3)- (ACI), Di(1 H-imidazol-1-yl-2,4,5-d3)methanone, N,N-Carbonyldiimidazole-d6, 1,1'-Carbonyldiimidazole-D6, 1,1'-Carbonyldiimidazole-d6
InChI formel InChI=1 S/C7H6N4O/c12-7(10-3-1-8-5-10)11-4-2-9-6-11/h1-6 H/i1D,2 D,3 D,4 D,5 D,6 D
SMIL [2 H]c1nc([2 H])n(C(=O)n2c([2 H])nc([2 H])c2[2 H])c1[2 H]
IUPAC navn bis(2,4,5-trideuterioimidazol-1-yl)methanone
Molekylvægt (g/mol) 168.1857
Formel vægt 168.0918 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity