Få mere at vide
1,1-Bis(4-chlorophenyl)ethane, Mikromol™
Artikelnummer. 17991236
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991236 100 mg
17951896 25 mg
30461717 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17991236 Leverandør Mikromol Leverandørnr. MM2513.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1-Bis(4-chlorophenyl)ethane
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1-Bis(4-chlorophenyl)ethane
CAS 4-4-3547
Analyt- eller komponentnavne 1,1-Bis(4-chlorophenyl)ethane
Forsendelsestilstand Room Temperature
Molekylær formel C14 H12 Cl2
Certificeringer/compliance ISO 17025
Synonym 1,1-Bis(4-chlorophenyl)ethane
InChI formel InChI=1S/C14H12Cl2/c1-10(11-2-6-13(15)7-3-11)12-4-8-14(16)9-5-12/h2-10H,1H3
SMIL CC(c1ccc(Cl)cc1)c2ccc(Cl)cc2
IUPAC navn 1-chloro-4-[1-(4-chlorophenyl)ethyl]benzene
Molekylvægt (g/mol) 251.15
Mængde 100 mg
Formel vægt 250.0316 g/mol