missing translation for 'onlineSavingsMsg'
Få mere at vide

[1,1'-Biphenyl]-4,4'-diol, 97%, BLD Pharm™

Artikelnummer. 31335084
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31335084 10 g
31325294 500 g
31322336 100 g
31318095 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31335084 Leverandør BLD Pharm Leverandørnr. BD1377510g
 Begrænset vare

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale [1,1'-Biphenyl]-4,4'-diol
CAS 92-88-6
Molekylær formel C12H10O2
MDL nummer MFCD00002348
Mængde 10 g
SMIL C2=C(C1=CC=C(O)C=C1)C=CC(=C2)O
Molekylvægt (g/mol) 186.21
Procent renhed 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.