missing translation for 'onlineSavingsMsg'
Få mere at vide

[1,1'-Biphenyl]-2’-tetrazolo-4-carboxylic acid, [1,1'-biphenyl]-2’-tetrazolo-4-ylmethyl ester (Losartan Impurity), TRC

Artikelnummer. 30328964
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328964 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30328964 Leverandør Toronto Research Chemicals Leverandørnr. TRCB3980001MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale [1,1'-Biphenyl]-2'-tetrazolo-4-carboxylic acid, [1,1'-biphenyl]-2'-tetrazolo-4-ylmethyl ester (Losartan Impurity)
CAS 1159977-10-2
Molekylær formel C28H20N8O2
Mængde 1 mg
InChI formel InChI=1S/C28H20N8O2/c37-28(21-15-13-20(14-16-21)23-6-2-4-8-25(23)27-31-35-36-32-27)38-17-18-9-11-19(12-10-18)22-5-1-3-7-24(22)26-29-33-34-30-26/h1-16H,17H2,(H,29,30,33,34)(H,31,32,35,36)
SMIL O=C(OCc1ccc(cc1)c2ccccc2c3nn[nH]n3)c4ccc(cc4)c5ccccc5c6nn[nH]n6
IUPAC navn [4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl 4-[2-(2H-tetrazol-5-yl)phenyl]benzoate
Molekylvægt (g/mol) 500.51
Formel vægt 500.1709
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.