missing translation for 'onlineSavingsMsg'
Få mere at vide

[1,1'-Bicyclohexyl]-1-carboxylic Acid, TRC

Artikelnummer. 30337507
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337507 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30337507 Leverandør Toronto Research Chemicals Leverandørnr. TRCB382435100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (1,1'-Bicyclohexyl)-1-carboxylic Acid
CAS 60263-54-9
Molekylær formel C13 H22 O2
Mængde 100 mg
Synonym [1,1'-Bicyclohexyl]-1-carboxylic acid, [Bicyclohexyl]-1-carboxylic acid (6CI,7CI), Bicyclohexyl-1-carboxylic acid, Dicycloverine EP impurity A
InChI formel InChI=1S/C13H22O2/c14-12(15)13(9-5-2-6-10-13)11-7-3-1-4-8-11/h11H,1-10H2,(H,14,15)
SMIL OC(=O)C1(CCCCC1)C2CCCCC2
IUPAC navn 1-cyclohexylcyclohexane-1-carboxylic acid
Molekylvægt (g/mol) 210.31
Formel vægt 210.162
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.