Learn More
1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone, 97%, Thermo Scientific™
Catalog Number p-4510287
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
1131-87-9
C10H7BrOS
255.13
MFCD07368508
NTQPGUCRHJHYIA-UHFFFAOYSA-N
1-1-benzothiophen-5-yl-2-bromo-1-ethanone, 1-1-benzothiophen-5-yl-2-bromoethanone, 1-benzo b thiophen-5-yl-2-bromoethan-1-one, 5-bromoacetylbenzo b thiophene, 5-bromoacetyl-benzo b thiophene, 5-bromoacetyl benzo b thiophene, ethanone,1-benzo b thien-5-yl-2-bromo, 1-benzo b thiophen-5-yl-2-bromoethanone, 1-1-benzothiophen-5-yl-2-bromoethan-1-one
7060546
1-(1-benzothiophen-5-yl)-2-bromoethanone
BrCC(=O)C1=CC=C2SC=CC2=C1
1131-87-9
C10H7BrOS
255.13
MFCD07368508
NTQPGUCRHJHYIA-UHFFFAOYSA-N
1-1-benzothiophen-5-yl-2-bromo-1-ethanone, 1-1-benzothiophen-5-yl-2-bromoethanone, 1-benzo b thiophen-5-yl-2-bromoethan-1-one, 5-bromoacetylbenzo b thiophene, 5-bromoacetyl-benzo b thiophene, 5-bromoacetyl benzo b thiophene, ethanone,1-benzo b thien-5-yl-2-bromo, 1-benzo b thiophen-5-yl-2-bromoethanone, 1-1-benzothiophen-5-yl-2-bromoethan-1-one
7060546
1-(1-benzothiophen-5-yl)-2-bromoethanone
BrCC(=O)C1=CC=C2SC=CC2=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone