Få mere at vide
1-[1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol Hydrochloride(rac N-Desmethyl Venlafaxine Hydrochloride), TRC
Artikelnummer. 30012769
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012769 25 mg
30011653 50 mg
17972915 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012769 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26625025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93413-90-2
Molekylær formel C16 H25 N O2 . Cl H
Molekylvægt (g/mol) 299.84
Synonym 1-[(1RS)-1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol Hydrochloride, Venlafaxine Hydrochloride Imp. D (EP) as Hydrochloride, N-Desmethylvenlafaxine Hydrochloride
IUPAC navn 1-[1-(4-methoxyphenyl)-2-(methylamino)ethyl]cyclohexan-1-ol;hydrochloride
SMIL Cl.CNCC(c1ccc(OC)cc1)C2(O)CCCCC2
Tekniske data
Kemisk navn eller materiale N-Desmethylvenlafaxine Hydrochloride
Mængde 25 mg
InChI formel InChI=1S/C16H25NO2.ClH/c1-17-12-15(16(18)10-4-3-5-11-16)13-6-8-14(19-2)9-7-13;/h6-9,15,17-18H,3-5,10-12H2,1-2H3;1H
Formel vægt 299.1652