Få mere at vide
1,1,3,3,5,5-Hexamethyltrisiloxane-1,5-diol, TRC
Artikelnummer. 30138378
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138378 100 mg
30010206 25 mg
30004994 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138378 Leverandør Toronto Research Chemicals Leverandørnr. TRCH296725100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3663-50-1
Molekylær formel C6H20O4Si3
Molekylvægt (g/mol) 240.48
Synonym 1,5-Dihydroxyhexamethyltrisiloxane, Bis[(hydroxydimethylsilyl)oxy]dimethylsilane, Hexamethyltrisiloxane-1,5-diol, 1,1,3,3,5,5-Hexamethyl-1,5-trisiloxanediol, Hexamethyl-1,5-trisiloxanediol;
IUPAC navn bis[[hydroxy(dimethyl)silyl]oxy]-dimethylsilane
SMIL C[Si](C)(O)O[Si](C)(C)O[Si](C)(C)O
Tekniske data
Kemisk navn eller materiale 1,1,3,3,5,5-Hexamethyltrisiloxane-1,5-diol
Mængde 100 mg
InChI formel InChI=1S/C6H20O4Si3/c1-11(2,7)9-13(5,6)10-12(3,4)8/h7-8H,1-6H3
Formel vægt 240.0669