Få mere at vide
1,1,2-Trichloroethane-d3, CDN
Artikelnummer. 30318242
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318242 0.25 g
30312745 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318242 Leverandør CDN Leverandørnr. CDND24090.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1,2-Trichloroethane-d3
CAS 171086-93-4
Molekylær formel C2 D3 Cl3
Mængde 0.25 g
Synonym Ethane-1,1,2-d3, 1,2,2-trichloro- (9 CI), 1,1,2-Trichloroethane-d3, 1,1,2-Trichloroethane-D3
InChI formel InChI=1 S/C2H3Cl3/c3-1-2(4)5/h2H,1H2/i1D2,2 D
SMIL [2 H]C([2 H])(Cl)C([2 H])(Cl)Cl
IUPAC navn 1,1,2-trichloro-1,2,2-trideuterioethane
Molekylvægt (g/mol) 136.4227
Formel vægt 134.9489 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity