Få mere at vide
1,1,2,2-Tetrahydroperfluoro-1-tetradecanol, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,1,2,2-Tetrahydroperfluoro-1-tetradecanol |
| CAS | 39239-77-5 |
| Molekylær formel | C14 H5 F25 O |
| Mængde | 50 mg |
| Synonym | 1H,1H,2H,2H-Perfluorotetradecanol, 1-Tetradecanol','3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-pentacosafluoro- (9CI','ACI), 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Pentacosafluoro-1-tetradecanol (ACI), 2-(Perfluorododecyl)ethanol, 2-(Perfluorododecyl)ethyl alcohol, Perfluorododecylethanol, PFA 12, 1H,1H,2H,2H-Perfluorotetradecan-1-ol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Pentacosafluoro-1-tetradecanol, 2-(Perfluorododecyl)ethanol, 2-(Perfluorododecyl)ethyl Alcohol, PFA 12, Perfluorododecylethanol |
| InChI formel | InChI=1S/C14H5F25O/c15-3(16,1-2-40)4(17,18)5(19,20)6(21,22)7(23,24)8(25,26)9(27,28)10(29,30)11(31,32)12(33,34)13(35,36)14(37,38)39/h40H,1-2H2 |
| SMIL | OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC navn | 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-pentacosafluorotetradecan-1-ol |
| Molekylvægt (g/mol) | 664.15 |
| Formel vægt | 663.9941 |