Learn More
1,1,2,2-Tetrabromoethane, Sigma-Aldrich™
Product Code. 15836820
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity Unit Size
15836820 1 L 1 L
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15836820 Supplier Sigma Aldrich Supplier No. 1000341000
 Restricted Item
Forventet forsendelse: 19-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 79-27-6
Melting Point -1°C
Density 2.96 g/cm3 at 25°C
Boiling Point 239°C/1013 hPa (Decomposition)
MDL Number MFCD00000133
Quantity 1 L
Synonym 1,1,2,2-Tetrabromoethane, TBE, Muthmanns liquid, Acetylene tetrabromide, 1,1,2,2-Tetrabromoethane
Molecular Weight (g/mol) 345.65
Grade Research