Få mere at vide
1,1,2,2-Tetrabromoethane-d2, CDN
Artikelnummer. 30310616
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310616 0.25 g -
30308935 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310616 Leverandør CDN Leverandørnr. CDND82430.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1,2,2-Tetrabromoethane-D2
CAS 33685-55-1
Molekylær formel C2 D2 Br4
Mængde 0.25 g
Synonym Ethane-1,2-d2, 1,1,2,2-tetrabromo- (8 CI, 9 CI), Ethane-1,2-d2, tetrabromo- (6 CI), 1,1,2,2-Tetrabromoethane-d2, 1,1,2,2-Tetrabromoethane-D2, Tetrabromoethane-1,2-d2
InChI formel InChI=1 S/C2H2Br4/c3-1(4)2(5)6/h1-2 H/i1D,2 D
SMIL [2 H]C(Br)(Br)C([2 H])(Br)Br
IUPAC navn 1,1,2,2-tetrabromo-1,2-dideuterioethane
Molekylvægt (g/mol) 347.6656
Formel vægt 343.7016 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity