Få mere at vide
1,1,1-Trichloro-2-(2-chlorophenyl-d4)-2-(4-chlorophenyl-d4)ethane, CDN
Artikelnummer. 30323999
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323999 0.01 g
30311989 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323999 Leverandør CDN Leverandørnr. CDND63800.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4'-DDT D8
CAS 221899-88-3
Molekylær formel C14 2H8 H Cl5
Mængde 0.01 g
InChI formel InChI=1 S/C14H9Cl5/c15-10-7-5-9(6-8-10)13(14(17,18)19)11-3-1-2-4-12(11)16/h1-8,13 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(c2c([2 H])c([2 H])c(Cl)c([2 H])c2[2 H])C(Cl)(Cl)Cl)c(Cl)c1[2 H]
IUPAC navn 1-chloro-2,3,4,5-tetradeuterio-6-[2,2,2-trichloro-1-(4-chloro-2,3,5,6-tetradeuteriophenyl)ethyl]benzene
Molekylvægt (g/mol) 362.54
Formel vægt 359.9649 g/mol
Procent renhed 98 atom % D, min 95% Chemical Purity