Få mere at vide
1,1,1-Trichloro-2-(2-chlorophenyl-d4)-2-(4-chlorophenyl-d4)ethane, CDN
Artikelnummer. 30311989
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311989 0.005 g
30323999 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311989 Leverandør CDN Leverandørnr. CDND63800.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Especificaciones
Kemisk navn eller materiale 2,4'-DDT D8
CAS 221899-88-3
Molekylær formel C14 2H8 H Cl5
Mængde 0.005 g
InChI formel InChI=1 S/C14H9Cl5/c15-10-7-5-9(6-8-10)13(14(17,18)19)11-3-1-2-4-12(11)16/h1-8,13 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(c2c([2 H])c([2 H])c(Cl)c([2 H])c2[2 H])C(Cl)(Cl)Cl)c(Cl)c1[2 H]
IUPAC navn 1-chloro-2,3,4,5-tetradeuterio-6-[2,2,2-trichloro-1-(4-chloro-2,3,5,6-tetradeuteriophenyl)ethyl]benzene
Molekylvægt (g/mol) 362.54
Formel vægt 359.9649 g/mol
Procent renhed 98 atom % D, min 95% Chemical Purity
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.