Få mere at vide
1,1'-[(1-Methylethyl)imino]bis[3-(naphthalen-1-yloxy)propan-2-ol] Hydrochloride (Tertiary Amine Derivative Hydrochloride), Mikromol™
Beskrivelse
1,1'-[(1-Methylethyl)imino]bis[3-(naphthalen-1-yloxy)propan-2-ol] Hydrochloride (Tertiary Amine Derivative Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,1'-[(1-Methylethyl)imino]bis[3-(naphthalen-1-yloxy)propan-2-ol] Hydrochloride |
| CAS | 2172495-51-9 |
| Analyt- eller komponentnavne | 1,1'-[(1-Methylethyl)imino]bis[3-(naphthalen-1-yloxy)propan-2-ol] Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C29 H33 N O4 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Propranolol Hydrochloride Imp. B (EP), Propranolol Imp. B (EP), 1,1'-[(1-Methylethyl)imino]bis[3-(naphthalen-1-yloxy)propan-2-ol] Hydrochloride, Propranolol Hydrochloride Imp. B (EP) as Hydrochloride, Tertiary Amine Derivative Hydrochloride of Propranolol, Propranolol Hydrochloride Impurity B as Hydrochloride, Propranolol Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C29H33NO4.ClH/c1-21(2)30(17-24(31)19-33-28-15-7-11-22-9-3-5-13-26(22)28)18-25(32)20-34-29-16-8-12-23-10-4-6-14-27(23)29;/h3-16,21,24-25,31-32H,17-20H2,1-2H3;1H |
| SMIL | Cl.CC(C)N(CC(O)COc1cccc2ccccc12)CC(O)COc3cccc4ccccc34 |
| IUPAC navn | 1-[(2-hydroxy-3-naphthalen-1-yloxypropyl)-propan-2-ylamino]-3-naphthalen-1-yloxypropan-2-ol;hydrochloride |
| Vis mere |