Få mere at vide
1,1'-[(1-Methylethyl)imino]bis[3-[4-(2-methoxyethyl)phenoxy]propan-2-ol], Mikromol™
Artikelnummer. 30049757
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30049757 100 mg
1 options
Artikelnummer. 30049757 Leverandør Mikromol Leverandørnr. MM0027.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1'-[(1-Methylethyl)imino]bis[3-[4-(2-methoxyethyl)phenoxy]propan-2-ol]
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1'-[(1-Methylethyl)imino]bis[3-[4-(2-methoxyethyl)phenoxy]propan-2-ol]
CAS 154784-36-8
Analyt- eller komponentnavne 1,1'-[(1-Methylethyl)imino]bis[3-[4-(2-methoxyethyl)phenoxy]propan-2-ol]
Forsendelsestilstand Room Temperature
Molekylær formel C27 H41 N O6
Certificeringer/compliance ISO 9001
Synonym 1,1'-[(1-Methylethyl)imino]bis-[3-[4-(2-methoxyethyl)phenoxy]propan-2-ol], Metoprolol Succinate Imp. O (EP), Metoprolol Tartrate Imp. O (EP), Metoprolol Imp. O (EP), Metoprolol USP Related Compound D, Metoprolol USP RC D, Metoprolol Related Compound D, Metoprolol Succinate Impurity O, Metoprolol Tartrate Impurity O, Metoprolol Impurity O, Metoprolol Impurity O
InChI formel InChI=1S/C27H41NO6/c1-21(2)28(17-24(29)19-33-26-9-5-22(6-10-26)13-15-31-3)18-25(30)20-34-27-11-7-23(8-12-27)14-16-32-4/h5-12,21,24-25,29-30H,13-20H2,1-4H3
SMIL COCCc1ccc(OCC(O)CN(CC(O)COc2ccc(CCOC)cc2)C(C)C)cc1
IUPAC navn 1-[[2-hydroxy-3-[4-(2-methoxyethyl)phenoxy]propyl]-propan-2-ylamino]-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
Molekylvægt (g/mol) 475.62
Mængde 100 mg
Formel vægt 475.2934 g/mol