Få mere at vide
1,1'-[(1-Methylethyl)imino]bis[3-[4-(2-methoxyethyl)phenoxy]propan-2-ol] Hydrochloride, Mikromol™
Beskrivelse
1,1'-[(1-Methylethyl)imino]bis[3-[4-(2-methoxyethyl)phenoxy]propan-2-ol] Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (+/-)N,N-bis-(2-Hydroxy-3-(4-(2-methoxyethyl)phenoxy)propyl)(1-methylethyl)amine hydrochloride |
| CAS | 1486464-40-7 |
| Analyt- eller komponentnavne | (+/-)N,N-bis-(2-Hydroxy-3-(4-(2-methoxyethyl)phenoxy)propyl)(1-methylethyl)amine hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C27H41NO6.ClH |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | Metoprolol Related Compound D, Metoprolol Related Compound D as Hydrochloride, Metoprolol Succinate Impurity O as Hydrochloride, Metoprolol Tartrate Impurity O as Hydrochloride, Metoprolol Impurity O as Hydrochloride, Metoprolol Impurity O as Hydrochloride |
| InChI formel | InChI=1S/C27H41NO6.ClH/c1-21(2)28(17-24(29)19-33-26-9-5-22(6-10-26)13-15-31-3)18-25(30)20-34-27-11-7-23(8-12-27)14-16-32-4;/h5-12,21,24-25,29-30H,13-20H2,1-4H3;1H |
| SMIL | Cl.COCCc1ccc(OCC(O)CN(CC(O)COc2ccc(CCOC)cc2)C(C)C)cc1 |
| IUPAC navn | 1-[[2-hydroxy-3-[4-(2-methoxyethyl)phenoxy]propyl]-isopropyl-amino]-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol;hydrochloride |
| Vis mere |