Få mere at vide
1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol], Mikromol™
Artikelnummer. 30051109
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051109 100 mg
30050926 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051109 Leverandør Mikromol Leverandørnr. MM0082.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol]
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol]
CAS 130115-65-0
Analyt- eller komponentnavne 1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol]
Forsendelsestilstand Room Temperature
Molekylær formel C25 H31 N3 O4
Certificeringer/compliance ISO 9001
Synonym 1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol], Pindolol Imp. C (EP), Pindolol Impurity C
InChI formel InChI=1S/C25H31N3O4/c1-17(2)28(13-18(29)15-31-24-7-3-5-22-20(24)9-11-26-22)14-19(30)16-32-25-8-4-6-23-21(25)10-12-27-23/h3-12,17-19,26-27,29-30H,13-16H2,1-2H3
SMIL CC(C)N(CC(O)COc1cccc2[nH]ccc12)CC(O)COc3cccc4[nH]ccc34
IUPAC navn 1-[[2-hydroxy-3-(1H-indol-4-yloxy)propyl]-propan-2-ylamino]-3-(1H-indol-4-yloxy)propan-2-ol
Molekylvægt (g/mol) 437.53
Mængde 100 mg
Formel vægt 437.2315 g/mol