missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroacetone hydrate, 98%, Thermo Scientific™

Catalog Number p-7077228
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10057-27-9
C3F6O
166.02
MFCD00149063
VBZWSGALLODQNC-UHFFFAOYSA-N
hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
36719
1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
FC(F)(F)C(=O)C(F)(F)F
This item is not returnable. View return policy
10057-27-9
C3F6O
166.02
MFCD00149063
VBZWSGALLODQNC-UHFFFAOYSA-N
hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
36719
1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
FC(F)(F)C(=O)C(F)(F)F
Melting Point 18.0°C to 21.0°C
Density 1.5800g/mL
Boiling Point 105.0°C to 108.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.58
Solubility Information Solubility in water: soluble.
Formula Weight 166.02
Percent Purity 98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone hydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexafluoroacetone hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'