Få mere at vide
1,1,1,3,3,3-Hexafluor-2-methoxypropan, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,1,1,3,3,3-hexafluoro-2-methoxy-propane |
| CAS | 13171-18-1 |
| Molekylær formel | C4 H4 F6 O |
| Mængde | 100 mg |
| Synonym | 1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Propane, 1,1,1,3,3,3-hexafluoro-2-methoxy- (9CI, ACI), 1,1,1,3,3,3-Hexafluoro-2-methoxypropane (ACI), Ether, methyl 2,2,2-trifluoro-1-(trifluoromethyl)ethyl (8CI), 1,1,1,3,3,3-Hexafluoroisopropyl methyl ether, 1-(Trifluoromethyl)-2,2,2-trifluoroethyl methyl ether, 1-Methoxy-1-trifluoromethyl-2,2,2-trifluoroethane, 356mmzEβγ, Bis(trifluoromethyl)methyl methyl ether, Hexafluoroisopropyl methyl ether, HFE 356mmz, Iso-flurothyl, Isoindoklon, Methyl 1-(trifluoromethyl)-2,2,2-trifluoroethyl ether |
| InChI formel | InChI=1S/C4H4F6O/c1-11-2(3(5,6)7)4(8,9)10/h2H,1H3 |
| SMIL | COC(C(F)(F)F)C(F)(F)F |
| IUPAC navn | 1,1,1,3,3,3-hexafluor-2-methoxypropan |
| Molekylvægt (g/mol) | 182.06 |
| Formel vægt | 182.0166 |