Få mere at vide
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-7-methyltridecane, TRC
Artikelnummer. 30453040
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30453040 100 mg
1 options
Artikelnummer. 30453040 Leverandør Toronto Research Chemicals Leverandørnr. TRCT907420100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-7-methyltridecane
CAS 2089686-13-3
Molekylær formel C14H17F13
Mængde 100 mg
InChI formel InChI=1S/C14H17F13/c1-3-4-5-6-7-8(2)9(15,16)10(17,18)11(19,20)12(21,22)13(23,24)14(25,26)27/h8H,3-7H2,1-2H3
SMIL FC(F)(C(F)(C(F)(C(F)(C(F)(C(F)(C(CCCCCC)C)F)F)F)F)F)F
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-7-methyltridecane
Molekylvægt (g/mol) 432.27
Formel vægt 432.11