Learn More
1,1',1'',1'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-2-ol), 98%, BLD Pharm™
Product Code. 31029698
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31029698 500 g
31023615 25 g
31025626 1 kg
31015575 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31029698 Supplier BLD Pharm Supplier No. BD141769500g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 102-60-3
Molecular Formula C14H32N2O4
MDL Number MFCD00004534
Quantity 500 g
SMILES CC(O)CN(CCN(CC(C)O)CC(C)O)CC(C)O
Molecular Weight (g/mol) 292.41
Percent Purity 98%
Chemical Name or Material 1,1',1'',1'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-2-ol)