missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Orai1 Antibody (Amgen patent anti-ORAI1) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-27962JF669
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Orai1 Monoclonal antibody specifically detects Orai1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| Orai1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1 | |
| Orai1 | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 84876 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Amgen patent anti-ORAI1 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion