missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Fascin Antibody (FSCN1/7209) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20939JF669
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Fascin Monoclonal antibody specifically detects Fascin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
| Fascin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin | |
| Recombinant fragment (around aa350-400) of human Fascin protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 6624 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| FSCN1/7209 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion