missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals ZNF254 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00009534-B01P
Additional Details : Weight : 0.00970kg
Description
ZNF254 Polyclonal antibody specifically detects ZNF254 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
ZNF254 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
AAH43147.2 | |
Mouse | |
Protein G purified | |
RUO | |
9534 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry | |
Unconjugated | |
PBS (pH 7.4) | |
BA526D8.4, FLJ33884, KRAB box containing C2H2 type zinc finger bA526D8.4, MGC133033, MGC133034, zinc finger protein 484 | |
ZNF254 (AAH43147.2, 1 a.a. - 659 a.a.) full-length human protein. MPGPPRSLEMGLLTFRDVAIEFSLEEWQHLDIAQQNLYRNVMLENYRNLAFLGIAVSKPDLITCLEQGKEPWNMKRHEMVDEPPGMCPHFAQGLWPEQGMEDSFQKAILRRYGKYGHENLQLRKGCKSVDEYKVNKEGYNGLNQCFTTAQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKSYKCKECGKTFNWSSTLTNHRKIYTEEKPYKCEEYNKSPKQLSTLTTHEIIHAGEKLYKCEECGEAFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTEHKKIHTRKKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKAFSQSSTLTTHKIIHTGEKRYKCLECGKAFKQLSTLTTHKIIHVGEKLYKCEECGKGFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTKHKRIHTREKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKSFSQSSTLTTHKIIHTGEKPYKCEECGKAFNWSSTLTKHNIIHTEEKPYKCEKCGKAFKQSSILTNHKRIHTGEKPYKCEECGKSFNRSSTFTKHKVIHTGVKPYKCEECGKAFFWSSTLTKHKRIHTGEQPYKWEKFGKAFNRSSHLTTDKITHWREILQV | |
0.05 mg | |
Primary | |
Human | |
Purified |