missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals GOSR1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00009527-B01P
Additional Details : Weight : 0.00970kg
Description
GOSR1 Polyclonal antibody specifically detects GOSR1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
GOSR1 | |
Polyclonal | |
Western Blot 1:500, Immunocytochemistry/ Immunofluorescence | |
NP_004862.1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
Unconjugated | |
PBS (pH 7.4) | |
golgi integral membrane protein 2, golgi SNAP receptor complex member 1,28 kDa cis-Golgi SNARE p28, Golgi SNARE 28 kDa, GOLIM2, GOS28cis-golgi SNARE, GOS-28GOS28/P28, GS2828 kDa Golgi SNARE protein, P28 | |
GOSR1 (NP_004862.1, 1 a.a. - 250 a.a.) full-length human protein. MAAGTSSYWEDLRKQARQLENELDLKLVSFSKLCTSYSHSSTRDGRRDRYSSDTTPLLNGSSQDRMFETMAIEIEQLLARLTGVNDKMAEYTNSAGVPSLNAALMHTLQRHRDILQDYTHEFHKTKANFMAIRERENLMGSVRKDIESYKSGSGVNNRRTELFLKEHDHLRNSDRLIEETISIAMATKENMTSQRGMLKSIHSKMNTLANRFPAVNSLIQRINLRKRRDSLILGGVIGICTILLLLYAFH | |
0.05 mg | |
Membrane Trafficking and Chaperones | |
9527 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |