missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals TTC11 Antibody (3G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00051024-M02
Additional Details : Weight : 0.00970kg
Description
TTC11 Monoclonal antibody specifically detects TTC11 in Human samples. It is validated for Western Blot, ELISASpecifications
TTC11 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
FIS1 (AAH03540.1, 1 a.a. ~ 152 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEAVLNELVSVEDLLKFEKKFQSEKAAGSVSKSTQFEYAWCLVRSKYNDDIRKGIVLLEELLPKGSKEEQRDYVFYLAVGNYRLKEYEKALKYVRGLLQTEPQNNQAKELERLIDKAMKKDGLVGMAIVGGMALGVAGLAGLIGLAVSKSKS | |
0.1 mg | |
Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
51024 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, ELISA | |
3G6 | |
Western Blot 1:500, ELISA | |
AAH03540 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |