missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals TRAPPC4 Antibody (2D10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00051399-M01
Additional Details : Weight : 0.00970kg
Description
TRAPPC4 Monoclonal antibody specifically detects TRAPPC4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISASpecifications
TRAPPC4 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
Hematopoietic stem/progenitor cell protein 172, PTD009, SBDNHSPC172, Synbindin, trafficking protein particle complex 4, trafficking protein particle complex subunit 4, TRS23, TRS23 homolog | |
TRAPPC4 (AAH10866, 1 a.a. ~ 219 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAIFSVYVVNKAGGLIYQLDSYAPRAEAEKTFSYPLDLLLKLHDERVLVAFGQRDGIRVGHAVLAINGMDVNGRYTADGKEVLEYLGNPANYPVSIRFGRPRLTSNEKLMLASMFHSLFAIGSQLSPEQGSSGIEMLETDTFKLHCYQTLTGIKFVVLADPRQAGIDSLLRKIYEIYSDFALKNPFYSLEMPIRCELFDQNLKLALEVAEKAGTFGPGS | |
0.1 mg | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA | |
2D10 | |
Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
AAH10866 | |
Mouse | |
IgG purified | |
RUO | |
51399 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |