missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals RAB6A Antibody (3G3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00005870-M01
Additional Details : Weight : 0.00970kg
Description
RAB6A Monoclonal antibody specifically detects RAB6A in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RAB6A | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
GTP-binding protein RAB6A', GTP-binding protein RAB6B, Oncogene RAB6, Rab GTPase, Rab-6, RAB6, member RAS oncogene family, RAB6A', Rab6A variant 3, RAB6A, member RAS oncogene family, RAB6B, RAB6rab-6, ras-related protein Rab-6A | |
RAB6A (AAH03617, 109 a.a. ~ 208 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DDVRTERGSDVIIMLVGNKTDLADKRQVSIEEGERKAKELNVMFIETSAKAGYNVKQLFRRVAAALPGMESTQDRSREDMIDIKLEKPQEQPVSEGGCSC | |
0.1 mg | |
Signal Transduction | |
5870 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3G3 | |
Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AAH03617 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |