missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals PMCA1 Antibody (3E2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00000490-M01
Additional Details : Weight : 0.00970kg
Description
PMCA1 Monoclonal antibody specifically detects PMCA1 in Human samples. It is validated for Western Blot, ELISA, ELISASpecifications
PMCA1 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
ATPase, Ca++ transporting, plasma membrane 1, EC 3.6.3, EC 3.6.3.8, plasma membrane calcium ATPase, Plasma membrane calcium ATPase isoform 1, plasma membrane calcium pump, Plasma membrane calcium pump isoform 1, plasma membrane calcium-ATPase, PMCA1kb, PMCA1plasma membrane calcium-transporting ATPase 1 | |
ATP2B1 (NP_001673, 1 a.a. ~ 97 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGDMANNSVAYSGVKNSLKEANHDGDFGITLAELRALMELRSTDALRKIQESYGDVYGICTKLKTSPNEGLSGNPADLERREAVFGKNFIPPKKPKT | |
0.1 mg | |
Cellular Markers | |
490 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA, Sandwich ELISA | |
3E2 | |
Western Blot 1:500, ELISA, Sandwich ELISA | |
NP_001673 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |