missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals PDLIM5 Antibody (3E11-F6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00010611-M01
Additional Details : Weight : 0.00970kg
Description
PDLIM5 Monoclonal antibody specifically detects PDLIM5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISASpecifications
PDLIM5 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5 | |
PDLIM5 (AAH08741, 1 a.a. ~ 596 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSNYSVSLVGPAPWGFRLQGGKDFNMPLTISSLKDGGKAAQANVRIGDVVLSIDGINAQGMTHLEAQNKIKGCTGSLNMTLQRASAAPKPEPVPVQKGEPKEVVKPVPITSPAVSKVTSTNNMAYNKAPRPFGSVSSPKVTSIPSPSSAFTPAHATTSSHASPSPVAAVTPPLFAASGLHANANLSADQSPSALSAGKTAVNVPRQPTVTSVCSETSQELAEGQRRGSQGDSKQQNGPPRKHIVERYTEFYHVPTHSDASKKRLIEDTEDWRPRTGTTQSRSFRILAQITGTEHLKESEADNTKKANNSQEPSPQLASSVASTRSMPESLDSPTSGRPGVTSLTTAAAFKPVGSTGVIKSPSWQRPNQGVPSTGRISNSAAYSGSVAPANSALGQTQPSDQDTLVQRAEHIPAGKRTPMCAHCNQVIRGPFLVALGKSWHPEEFNCAHCKNTMAYIGFVEEKGALYCELCYEKFFAPECGRCQRKILGEVINALKQTWHVSCFVCVACGKPIRNNVFHLEDGEPYCETDYYALFGTICHGCEFPIEAGDMFLEALGYTWHDTCFVCSVCCESLEGQTFFSKKDKPLCKKHAHSVNF | |
0.1 mg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA | |
3E11-F6 | |
Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
AAH08741 | |
Mouse | |
IgG purified | |
RUO | |
10611 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |