missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals MPG Antibody (4B12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00004350-M08
Additional Details : Weight : 0.00970kg
Description
MPG Monoclonal antibody specifically detects MPG in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISASpecifications
MPG | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on16p, 3-alkyladenine DNA glycosylase, 3-methyladenine DNA glycosidase, AAG, ADPG, ANPG, APNG, CRA36.1, CRA36.1 (3-methyl-adenine DNA glycosylase), DNA-3-methyladenine glycosylase, EC 3.2.2.21, MDGalkyladenine DNA glycosylase, Mid1, N-methylpurine-DNA glycosylase, MPG, N-methylpurine-DNA glycosylasePIG16, PIG11, proliferation-inducing protein 11, proliferation-inducing protein 16 | |
MPG (AAH14991, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPARSGAQFCRRMGQKKQRPARAGQPHSSSDAAQAPAEQPHSSSDAAQAPCPRERCLGPPTTPGPYRSIYFSSPKGHLTRLGLEFFDQPA | |
0.1 mg | |
Direct Reversal of DNA Damage | |
4350 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA | |
4B12 | |
Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
AAH14991 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |